Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBC1D2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$428.82 - $684.18

Specifications

Antigen TBC1D2
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB159915
SDP
View Documents
Novus Biologicals
NBP31794225UL
25 μg
Each for $428.82
Only null left
Add to Cart
 
NB159914
SDP
View Documents
Novus Biologicals
NBP317942100UL
100 μg
Each for $684.18
Only null left
Add to Cart
 
Description

Description

TBC1D2 Polyclonal antibody specifically detects TBC1D2 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

TBC1D2
Immunofluorescence
Unconjugated
Rabbit
Human
Armus, FLJ10702, FLJ16244, FLJ42782, PARIS1member 2A, PP8997, TBC1 domain family, member 2
This antibody was developed against Recombinant Protein corresponding to amino acids: VAEKEKALLTKCAYLQARNCQVESKYLAGLRRLQEALGDEASECSELLRQLVQEALQWEAGEASSDSIELSPISKYDEYGFLTVPDYEVEDLKLL
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Polyclonal
Purified
RUO
PBS, pH 7.2, 40% glycerol
55357
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.