Learn More
Medchemexpress LLC HY-100975 50mg Medchemexpress, TB5 CAS:948841-07-4 Purity:>98%
Supplier: Medchemexpress LLC HY10097550MG
Medchemexpress, HY-100975 50mg TB5 CAS:948841-07-4 TB5 is a potent, selective and reversible inhibitor of hMAO-B with a Ki value of 0.11±0.01 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.