Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Exogenous Internal Positive Control Reagents
GREENER_CHOICE

Catalog No. p-7083360
Encompass_Preferred
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions

Catalog No. 43-083-21

Supplier: Applied Biosystems™ 4308321

Only null left
Add to Cart
Edge
Add to Cart

TaqMan™ Exogenous Internal Positive Control Reagents contain a pre-optimized internal positive control (IPC) with pre-designed primers and TaqMan™ probe.

  • Applied Biosystems™ TaqMan™ Exogenous Internal Positive Control Reagents contain a pre-optimized internal positive control (IPC) with pre-designed primers and TaqMan™ probe
  • The IPC can be spiked into samples to distinguish true target negatives from PCR inhibition
  • This kit allows users to amplify a low-copy target DNA in the same tube with the IPC
  • Although the target and IPC DNAs may differ in initial copy number, the concentration of the IPC primers in the PCR reaction is limiting so that the amplification efficiency of the target reaction is not compromised
  • Pre-designed primers and TaqMan™ probe eliminate assay design
  • Rapid assay development guidelines can minimize optimization time Separate components provide flexibility in assay set-up
  • Reagents may be stored at 2°C to 4°C if they are to be consumed within one month
  • Otherwise, store at -20°C to -25°C

Note: This is not a quantitative kit and should not be used for normalization Instead, this kit is designed to distinguish types of negative results: a negative call for the target sequence and a positive call for the IPC indicate that no target sequence is present, and a negative call for the target sequence and negative call for the IPC suggest PCR inhibition.

Order Info

Note: This is not a quantitative kit and should not be used for normalization. Instead, this kit is designed to distinguish types of negative results: a negative call for the target sequence and a positive call for the IPC indicate that no target sequence

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 5 of the following:
• 1 ml 10X Exo IPC Mix
• 120 μl 10X Exo IPC Block
• 200 μl 50X Exo IPC DNA

Store at -20°C to -25°C. Reagents may be stored at 2–4°C if they are to be consumed within one month.
Format Tube(s)
GC-Rich PCR Performance Low
PCR Method qPCR
Reaction Speed Standard
Green Features Sustainable packaging
Internal Probe Modification TAMRA™ Quencher (3'), VIC™ (5')
Product Line TaqMan
Product Type Exogenous Internal Positive Control Reagent
Quantity 1,000 reactions
Sufficient For 1000 Reactions
No. of Reactions 1000 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.