Learn More
Lumiprobe TAMRA azide, 5-isomer, 1 mg
Supplier: Lumiprobe A7130
TAMRA (TMR, tetramethylrhodamine) azide, solid compound, and 10 mM solution in DMSO, labeling reagent for Click Chemistry. Pure 5-isomer. TAMRA is often used as FRET acceptor for FAM fluorophore.