Learn More
Clontech Labs 3P TALON® Metal Affinity Resin 100 ml
Supplier: Clontech Labs 3P 635503
Immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins under native or denaturing conditions.100 ml
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.