Learn More
Trajan Scientific SYR 2.5ML 2.5MDR-VLL-GT
Supplier: Trajan Scientific 008560
SGE Syringes with Valves, Volume: 2.5 mL, Borosilicate Glass, Autoclavable: Autoclavable, Connection Type: Removable, For Use With: For sample storage and transportation or to pressurize sample prior to injection, Termination: Luer-lock