Learn More
Kingfisher Biotech Inc Swine CCL2 (MCP-1) ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY0719S004
Encompass_Preferred
The Swine CCL2 (MCP-1) ELISA contains capture antibody protein standard and detection antibody for development of a Swine CCL2 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include CCL2 GDCF-2 HC11 HSMCR30 MCAF MCP-1 MCP1 SCYA2 SMC-CF C-C motif chemokine ligand 2. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5220