Learn More
SUV3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303055A
Description
The recommended shelf life for this product is 1 year from date of receipt.
ATPase and DNA/RNA helicase able to unwind DNA/DNA, DNA/RNA and RNA/RNA duplexes in the 5′-3′ direction. May protect cells from apoptosis.Specifications
| SUV3 | |
| Polyclonal | |
| Unconjugated | |
| SUPV3L1 | |
| SUV3, ATP-dependent RNA helicase SUPV3L1, mitochondrial, SUV3-like protein 1, suppressor of var1 3-like protein 1, suppressor of var1, 3-like 1(SUV3) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6832 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8IYB8 | |
| SUPV3L1 | |
| Between 686 and 736 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.