Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 6-0, PS-3, 18in, 12/Box
Supplier: Med Vet International 1665G
The Ethicon 1665G suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a PS-3 Precision Point reverse cutting needle and has 12 sutures in a box.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.