Learn More
Medchemexpress LLC HY-19285 100mg Medchemexpress, Sulfaclozine CAS:102-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19285100MG
Encompass_Preferred
Medchemexpress, HY-19285 100mg Sulfaclozine CAS:102-65-8 Sulfaclozine (Sulfachloropyrazine) is an efficacious sulphonamide derivative with antibacterial and anticoccidial effects. Sulfaclozine is commonly used for the treatment of various poultry diseases (particularly, collibacteriosis, fowl cholera and coccidiosis). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2111