Learn More
World Precision Instrument STX4 EVOM™ Electrode with Removable Blades for TEER in 6.5 mm Inserts
Supplier: World Precision Instrument EVMEL030301
Electrode for EVOM ® Manual with Replaceable Blades Ensures Stable Measurements
The STX4 electrode is the most recent version of chopstick electrode for the EVOM ® Manual (or the older version, EVOM3), and it is designed to facilitate measurements of voltage (potential difference: PD) and resistance (transepithelial electrical resistance: TEER) of cultured epithelia (cellular layer) grown onto permeable membrane inserts in 12- and 24-well tissue culture plates. STX4 can measure directly inside the well plates with 12 and 24 inserts. The STX4 is designed primarily for 24-well hanging inserts or transwells (e.g., Corning 3470), and the electrode¿s best performance can be seen in this format.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.