Learn More
New England Biolabs, Inc. Streptavidin – 1 mg
SDP
Supplier:  New England Biolabs, Inc. N7021S
Encompass
A 52.8 kDa non-glycosylated protein with a very high affinity for biotin. Due to its high affinity for biotin, it is used to bridge biotinylated probes and biotinylated enzymes.
Catalog No. 50-811-808