Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck CVA™ Calibration Verification Assessment/Linearity Kit
SDP

Catalog No. p-108958 Shop All Streck Laboratories Products
Click to view available options
Quantity:
5 x 3 mL
12 x 3 mL
13 x 3 mL
16 x 3 mL
18 x 3 mL
Intended Use:
CVA for RVC, HgB
CVA for WBC, Platelets, HgB
For Use With (Equipment):
ADVIA 120 and LH750
ADVIA 60 Hematology Analyzer
CELL DYN 3000
CELL DYN 3200
HemoCue Hematology Analyzer
This item is not returnable. View return policy

Catalog No. 11-716-320

Supplier: Streck 291504

This item is not returnable. View return policy

CVA (Calibration Verification Assessment) and CVA for CELL-DYN are assayed linearity control kits that test the upper and lower limits of three- and five-part differential hematology analyzers to determine their patient reportable range and linear performance.

  • Provides assay values for the most popular three- and five-part hematology instruments.
  • Customized kits with WBC, RBC, Hgb and platelet ranges for each instrument model.
  • Plastic vials with pierceable caps for analyzer autosampling.
  • 5-day open-vial stability, 120-day closed-vial stability.
  • Tests the extreme upper and lower limits of an instrument.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

For Use With (Equipment) HemoCue Hematology Analyzer
Type Linearity Control Kit
Includes WBC, platelet and RBC or Hemoglobin vials
Intended Use CVA for RVC, HgB
Level 5
Quantity 5 x 3 mL
Stability 5 Days Open, 120 Days Closed Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.