Learn More
Description
- Consists of four quaternary ammonium compounds formulated with a compatible detergent system
- Effective in hard water up to 400ppm hardness as CaCO3 and in the presence of 5% blood serum
- Provides broad spectrum germicidal activity; effective against influenza, HIV-1 virus, and Hepatitis B and C
- Contains no dyes or perfumes that could interfere with critical research studies or manufacturing processes
- Phosphate-free
- Used at a 1:256 dilution
Specifications
Specifications
| Form | Liquid |
| pH | 13 |
| Appearance | Clear |
| Certifications/Compliance | NOISH, OSHA |
| Composition | 13.02% Di-n-alkyl Dimethyl Ammonium Chloride, 8.68% N-alkyl Dimethyl Benzyl Ammonium Chloride, Ethyl |
| For Use With (Application) | Provides broad spectrum germicidal activity for simultaneous cleaning and disinfecting of washable, hard, non porous environmental surfaces such as floors, walls and processing equipment |
| Volume (English) | 1 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.