Learn More
Reprocell STEMOLECULE CHIR99021 (10MG)
Supplier: Reprocell 04000410
Size 2 mg (Cat. No. 04-0004) 10 mg (Cat. No. 04-0004-10) Alternate Name 6-[[2-[[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2 pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile Chemical Formula C22H18Cl2N8 Molecular Weight 465.34 CAS Number 252917-06-9 Purity Greater than 95% by HPLC analysis Formulation Off-white solid Solubility Reconstitute in DMSO to the desired concentration. For reconstitution instructions please reference product specifications sheet. Storage and Stability Store powder at 4°C protected from light. Following reconstitution, store aliquots at -20°C. Stock solutions are stable for 6 months when stored as directed. Quality Control The purity of CHIR99021 was determined by HPLC analysis. The accurate mass was determined by mass spectrometry. Cellular toxicity of CHIR99021 was tested on mouse embryonic stem cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.