Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America StatSpin™ LipoClear™ Tubes

Catalog No. p-179768
Click to view available options
Quantity:
0.5 mL (40/Pk.)
Type:
Tube
This item is not returnable. View return policy

Catalog No. 22-274915

Supplier: HemoCue America LC40

This item is not returnable. View return policy

Clear lipemic serum or plasma with any standard centrifuge: no ultracentrifuge or organic solvents needed

  • LipoClear tubes are pre-filled and ready-to-use with a nontoxic, non-carcinogenic reagent
  • Clearing time varies with centrifuge model (only 95 seconds with StatSpin-MP)
  • LipoClearPlus in 30mL size available for higher volume laboratories

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Format Tube
Type Tube
For Use With (Application) Blood collection
For Use With (Equipment) Tube rotor, can also be used in StatSpin LipoClear high speed centrifuge and other standard centrifuge
Quantity 0.5 mL (40/Pk.)
Sample Size 0.1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.