Learn More
Electron Microscopy Sciences Plastic Box, Hinged Lid Snap Lock, 1" Square x 5/8"
Supplier: Electron Microscopy Sciences 64310
The sqare boxes are molded from polystyrene for optimum clarity and rigidity. They have two equal dimensions for length and width. With hinged snap-lock lid.
Square: 1"
Height: 5/8"
Lid: 1/8"
Base: 1/2"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.