Learn More
Med Vet International Dynarex Non-Woven Sponge, Sterile 2'S, 2" X 2", 4 Ply, 1500/Case
Supplier: Med Vet International 3362
Non-woven sponges are ideal for general purpose cleaning and dressing of wounds. Ideal for wound dressing, prepping and scrubbing, general purpose cleansing Highly absorbent Less linting than standard gauze 3362-3364: Packaged in peel-down pouches for convenient, aseptic handling; Pouches packed in open trays for easy access
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.