Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Hydraulic Laboratory Pellet Press Accessories, Pellets

Catalog No. p-4909427
Click to view available options
Diameter (Metric):
13 mm
31 mm
35 mm
40 mm
Material:
Steel
Tungsten

Catalog No. 3613ST

Supplier: Cole-Parmer™ 3613ST

Only null left
Add to Cart
Add to Cart

Optimize performance of your Cole-Parmer™ hydraulic laboratory pellet press with these Pellets.

  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more.
  • Ensure top results from your Cole-Parmer™ hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components.
  • The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.

Specifications

Type Pellet
Diameter (English) 0.5 in.
Diameter (Metric) 13 mm
For Use With (Application) For Routine Pressing of Non-Abrasive Materials
For Use With (Equipment) For 3613 Evacuable Pellet Die Set
Material Steel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.