Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ Grinding Vial Set, Large Vial, PC Cylinder, Low-Chrome Steel End Plugs and Impactor
Click to view available options
Capacity:
1 to 50 mL (0.03 to 5.07 oz.)
Description
- Grinds one large sample at a time in the Large Freezer/Mill™
- Sample grinding capacity 5 g – 50 g.
- Specifically designed to grind electronic components for RoHS/WEEE compliance testing.
- End plugs and impactor are made of low-chrome ASTM 06 steel to eliminate chromium contamination during grinding.
- Withstands liquid nitrogen temperatures.
- Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples.
- Transparent plastic center cylinder allows visual checking of the grinding progress.
- low-chrome steel end plugs and impactor are autoclavable and reusable.
- Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent.
- Compatible with 6804 Large Extractor/Vial Opener.
Specifications
Specifications
| Capacity (English) | 0.03 to 5.07 oz. |
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | 6870 Freezer/Mill |
| For Use With (Application) | For Grinding Electronic Components |
| Material | Low-chrome Steel |
| Capacity | 1 to 50 mL (0.03 to 5.07 oz.) |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction