Learn More
Description
- Features QRS (Quick Release Swivel), which enables one-handed connection and release.
- Extend the service life of the breathing tube by using a breathing tube cover
- For maximum safety and comfort, make it a habit to check equipment regularly.
Specifications
Specifications
| Color | Black |
| For Use With (Application) | Metal repair, metal processing, welding, grinding |
| For Use With (Equipment) | Adflo™ Powered Air Purifying Respirator |
| Model | 70071735982 |
| Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.