Learn More
Medchemexpress LLC HY-10343 10mg , Sotrastaurin CAS:425637-18-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1034310MG
Encompass_Preferred
HY-10343 10mg Sotrastaurin CAS: 425637-18-9 Sotrastaurin (AEB071) is a potent and orally-active pan-PKC inhibitor, with Kis of 0.22 nM, 0.64 nM, 0.95 nM, 1.8 nM, 2.1 nM and 3.2 nM for PKCθ, PKCβ, PKCα, PKCη, PKCδ and PKCε, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0446