Learn More
Chemglass Life Sciences 500mL Solvent Distillation Still Head, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF070002
500mL Solvent Distillation Still Head, Airfree, Schlenk. Still head in 500mL capacity for use in re-purification of solvents under inert atmosphere. One-piece design of condenser and storage reservoir eliminates multiple connections usually associated with distillation setup. 0-4mm Chem-Vac™ Chem-Cap valves provide for grease-free, leakproof seals. Lower 3-way valve (C) allows for solvent return to flask, take-off or storage in graduated reservoir. Syringe access port (B) incorporates a 4mm PTFE stopcock with a 14/20 outer joint for use with CG-3022 septum stoppers permitting repeated syringe punctures or changes of the septum without breaching the integrity of the solvent. Valve (A) allows for isolation of the still from the inert gas source. Valve (D) permits evacuation and purging of the receiver flask. Uptake tube (F) is a 24/40 inner joint, while the takeoff connection (E) is a 14/20 inner joint. Design Suggested by Dr. Thomas Oberhauser.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.