Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphite dibasic pentahydrate, 98%

Catalog No. AC428502500
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle

Catalog No. AC428502500

Supplier: Thermo Scientific Chemicals 428502500

Only null left
Add to Cart

CAS: 13517-23-2 | HNa2O35H2O | 216.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13517-23-2
Molecular Formula HNa2O35H2O
Molecular Weight (g/mol) 216.02
Synonym sodium phosphite dibasic pentahydrate, di-sodium hydrogen phosphite, di-sodium hydrogen phosphite pentahydrate, disodium pentahydrate hydrogen phosphite, 2na.ho3p.5h2o, di-sodiumhydrogenphosphitepentahydrate, disodium hydrogenphosphite pentahydrate, etidronate disodium related compound a, united states pharmacopeia usp reference standard

Specifications

Quantity 250 g
Assay Percent Range 98%
Merck Index 15,8793
Solubility Information Solubility in water: soluble
Formula Weight 216.02
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Sodium phosphite dibasic pentahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.