Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate, dibasic dihydrate, 98%, extra pure

Catalog No. AC448150010
Click to view available options
Quantity:
1 kg
5 kg

Catalog No. AC448150010

Supplier: Thermo Scientific Chemicals 448150010

Only null left
Add to Cart

CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
IUPAC Name disodium dihydrate hydrogen phosphate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 243.0°C to 245.0°C
Quantity 1 kg
Assay Percent Range 98%
Linear Formula Na2HPO4·2 H2O
Merck Index 15, 8789
Solubility Information Solubility aq. soln.: 93 g/L(20°C).
Formula Weight 177.99
Grade Extra Pure
Chemical Name or Material Sodium phosphate, dibasic dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.