Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexachloropalladate(IV) hydrate, 99.9% (metals basis)

Catalog No. p-7048619
Click to view available options
Quantity:
1 g
5 g

Catalog No. AA1103903

Supplier: Thermo Scientific Chemicals 01103903

Only null left
Add to Cart

CAS: 53823-60-2 | Cl6Na2Pd | 365.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53823-60-2
Molecular Formula Cl6Na2Pd
Molecular Weight (g/mol) 365.10
MDL Number MFCD00014235
InChI Key OWJQEDIJRBKDJO-UHFFFAOYSA-H
IUPAC Name disodium hexachloropalladiumbis(ylium)
SMILES [Na+].[Na+].Cl[Pd++](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Brown to Red
Linear Formula Na2PdCl6·xH2O
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 365.10 (Anhydrous)
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Sodium hexachloropalladate(IV) hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 258-809-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.