Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Chloride Standard, 1000 ppm NaCl, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
| Boiling Point | 100°C |
| Melting Point | 0.0°C |
| CAS | 7732-18-5, 7647-14-5, 7647-14-5, 7732-18-5 |
| Color | Colorless |
| CAS Min % | 99.9%, 0.1% |
| Physical Form | Liquid |
| Quantity | 500 mL |
| CAS Max % | 0.1%, 99.9% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 1000 ppm |
| Show More |
Safety and Handling
| ShelfLife | 24 Months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction