Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Bromide (Granular/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
Specifications
Specifications
| Boiling Point | 1390°C |
| Melting Point | 755°C |
| Color | White |
| pH | 5 to 8.8 |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | ≥99 % |
| Formula Weight | 102.89 |
| Percent Purity | ≥99% |
| Grade | Certified ACS |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction