Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% solution in toluene (ca. 3.5M), AcroSeal™

Catalog No. AC430911000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

Catalog No. AC430911000

Supplier: Thermo Scientific Chemicals 430911000

Only null left
Add to Cart

CAS: 22722-98-1 | C6H16AlNaO4 | 202.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22722-98-1, 108-88-3
Molecular Formula C6H16AlNaO4
Molecular Weight (g/mol) 202.16
MDL Number MFCD00011631
Synonym Sodium dihydro-bis-(2-methoxyethoxy)aluminate in toluene

Specifications

Boiling Point 110.0°C
Color Amber to Colorless
Physical Form Viscous Liquid
Quantity 100 mL
Linear Formula [(CH3OCH2CH2O)2AlH2]Na
Flash Point 4°C
Merck Index 15,8702
Solubility Information Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers
Formula Weight 202.16
Percent Purity 68.5 to 71.5%
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.036
Density 1.0360g/mL
Chemical Name or Material Sodium bis(2-methoxyethoxy)Aluminum hydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May be fata
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 245-178-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.