Learn More
Quality Biological Inc SOC Medium, PK 10 X 10 mL
Supplier: Quality Biological Inc 340031671
SOC medium is a ready-to-use solution ¿ Useful in the final step of bacterial cell transformation to obtain maximum transformation efficiency of E. coli. ¿ Without magnesium chloride ¿ Used for the transformation of competent cells (E. coli). ¿ Used in the propagation of DH-alpha (of E. coli strains) primarily in the recovery step of Escherichia coli competent cell transformations, and to maximize the transformation efficiency of competent cells. ¿ Recommendation: Immediately before use, add 2M magnesium chloride to a final concentration of 20mM (10ml MgCl2/L). ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Autoclave Sterilized ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 41106200 ¿ Max Shelf Life: 24 months
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.