Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab Snap n' Digest Tuberculosis Collection Kit

Click to view available options
Volume (Metric):
75 mL
150 mL
Description
- Contains crushable NALC ampoules inside a safety straw inside a flexible bottle with alkali solution and TB phosphate buffer packets.
- Compatible with multiple automated TB systems
- Solution in bottle with unbroken ampoule is stable for 1 year without refrigeration
- 10 bottles/box
Available in 75 mL or 150 mL:
- 75 mL contains 1 NALC ampoule, 4 oz. bottle with 2 oz. of alkali solution, and 5 TB Phosphate Buffer packets
- 150 mL contains 2 NALC ampoules, 8 oz. bottle with 4 oz. of alkali solution, and 10 TB Phosphate TB Buffer packets
Specifications
Specifications
| Product Type | Collection and Transport System |
| Format | Tube |
| Sterility | Non-sterile |
| Quantity | 10 Bottles/Box |
| Volume (Metric) | 75 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction