Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Snap Circuits Sets
SDPSCI_ED

Catalog No. p-2366581
Encompass
Click to view available options
Product Type:
Snap Circuits 300 Experiment
Snap Circuits Jr. 100 Experiment

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S66934

Supplier: Elenco Electronics Inc SC300

Only null left
Add to Cart
Add to Cart

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

Instruction manuals

Create exciting experiments.

Enjoy hours of educational fun while building projects such as AM radios, burglar alarms, flashlights, doorbells and electronic games. Project components feature easy-snap assembly with no tools required.

  • Snap Circuits offers more than 300 project options; requires four AA batteries (not included)
  • Snap Circuits Jr. offers over 100 project options; requires two AA batteries (not included)

Specifications

Product Type Snap Circuits 300 Experiment
For Use With (Application) To build equipments such as AM radio, burglar alarm, flashlight, doorbell and electronic games
Includes Instruction manual
Weight (English) 3.1 lb.
Weight (Metric) 1.4 kg
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.