Learn More
ACROBiosystems Recombinant Protein;Human SIRP gamma / CD172g Protein, Fc Tag;HEK293;100UG;SIG,SIRPG,CD172g,SIRPB2,SIRP-gamma,SIRP-b2,SIRP-beta-2,SIRPB-2,Cripto-1
Supplier:  ACROBiosystems SIGH5253100UG
Encompass_Preferred
Recombinant Protein;Human SIRP gamma / CD172g Protein, Fc Tag;100UG;Human SIRP gamma, Fc Tag (SIG-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 29 - Ser 364 (Accession # AAH64532).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.7 kDa. The protein migrates as 68-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, His Tag (Cat. No. CD7-H5227) at 5 μg/mL (100 μL/well) can bind Human SIRP gamma, Fc Tag (Cat. No. SIG-H5253) with a linear range of 0.01-0.156 μg/mL (QC tested).
Catalog No. 50-165-8505
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.