Learn More
Strem, An Ascensus Company CAS# 13465-84-4. 10g. Silicon(IV) iodide (99.9%-Si). MFCD00054041
SDP
Supplier:  Strem, An Ascensus Company 93144710G
Encompass
CAS# 13465-84-4. 10g. Silicon(IV) iodide (99.9%-Si). MFCD00054041. Molecular Weight: 535.70. Molecular Formula: SiI4. Color/form: off-white pwdr. Strem# 93-1447. http://www.strem.com/catalog/v/93-1447/
Catalog No. 50-901-14130