Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, Grade 47 (10-18 Mesh/Certified ACS), Fisher Chemical™

Catalog No. p-33410 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
200 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. S161-500

Only null left
Add to Cart

Specifications

Chemical Name or Material Silica Gel
Color Blue
Physical Form Solid
Name Note Tel-Tale™ Desiccant, Indicating, 10-18 Mesh
CAS 7646-79-9, 7631-86-9
Material Silica Gel
Quantity 500 g
Identification Pass Test
Molecular Formula SiO2
MDL Number MFCD00011232
Mesh Size 10 to 18 Mesh
Packaging Poly Bottle
Suitability Pass Test
Grade Certified ACS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.