Learn More
Siemens Healthineers Urinalysis Reagent Test Strips
Catalog No. AM2179 Shop All Siemens Healthineers Products
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, pH, Protein, Urobilinogen
Blood
Blood Urea, Nitrogen
Blood, Glucose, Ketone, pH, Protein
Blood, Glucose, pH, Protein
Glucose, Protein
Urine Protein
Quantity:
100 Pack
25 Pack
50 Pack
This item is not returnable. View return policy
Catalog No. AM-2179
Supplier: Siemens Healthineers 10339953
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Single-use, plastic strips with multiple reagent test areas
  • Ready to use and entirely disposable
  • Dip and read visually for results
  • Some strips can be read instrumentally on Clinitek™ 50, 100, 200, 200+, 500, Advantus, and Status Family of Urine Chemistry analyzers
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Bilirubin, Blood, Glucose, Ketone, pH, Protein, Urobilinogen
Disposable Single-use
For Use With (Application) Urinalysis, in-vitro diagnostic use
Quantity 100 Pack
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.