Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGCG Rabbit anti-Human, Unconjugated, Clone: AbS34, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20069
Description
SGCG Monoclonal antibody specifically detects SGCG in Human samples. It is validated for IP, IHC, WB-Ce.Specifications
| SGCG | |
| Monoclonal | |
| Unconjugated | |
| SGCG | |
| Rabbit | |
| 100 μg | |
| 6445 | |
| Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
| IgG |
| Immunoprecipitation, Immunohistochemistry, Western Blot | |
| AbS34 | |
| sarcoglycan, gamma (35kDa dystrophin-associated glycoprotein) | |
| A4, DAGA4, DMDA, DMDA1, LGMD2C, MAM, MGC130048, SCARMD2, SCG3, TYPE | |
| A synthetic peptide corresponding to human SGCG | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction