Learn More
Aviva Systems Biology Corp SFN Antibody - N-terminal region (ARP54791_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54791_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SFN. Alternate names: YWHASApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-967