Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesamin 98.0+%, TCI America™
SDP

Catalog No. S0495100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0495100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0495100MG Supplier TCI America Supplier No. S0495100MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 607-80-7
Molecular Formula C20H18O6
Molecular Weight (g/mol) 354.358
MDL Number MFCD00216134
InChI Key PEYUIKBAABKQKQ-USWSZDBUSA-N
PubChem CID 44714838
IUPAC Name 5-[(3S,3aR,6aR)-3-(1,3-benzodioxol-5-yl)-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-6-yl]-1,3-benzodioxole
SMILES C1C2C(COC2C3=CC4=C(C=C3)OCO4)C(O1)C5=CC6=C(C=C5)OCO6

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 100 mg
Formula Weight 354.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Sesamin
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.