Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Secador™ 5.0 Auto-Desiccator Cabinet, Electric Non-Desiccant

Catalog No. 01257491 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
No. of Shelves:
3
This item is not returnable. View return policy

Catalog No. 01-257-491

Supplier: Bel-Art™ H420571003

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Desiccant-free cabinet provides dry storage for humidity-sensitive products such as laboratory samples, reagents, optical equipment, electronics, and 3D printing filament.

  • High-density foam gaskets to keep ambient humidity out and dry air inside
  • Large clear polycarbonate panels are durable and allow for easy viewing of internal contents
  • LCD shows the internal temperature and humidity level
  • Includes 3 adjustable and removable shelves
  • Electric models keeps internal humidity at 20-40%

Specifications

Autoclavable Not Autoclavable
Color White
Cabinet Style Front-loading
Material Polycarbonate
Product Type Desiccator
Door Style Hinged
No. of Shelves 3
Shelf Positions Adjustable
Dimensions (D x W x H) Interior 34 cm x 28 cm x 50cm
Width (English) 11.8 in.
Width (Metric) 30 cm
Height (English) 25 in.
Height (Metric) 63.5 cm
Depth (English) 13 in.
Depth (Metric) 34 cm
Voltage 100/120 V or 200/240 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.