Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl Crotonate 97.0+%, TCI America™
SDP

Catalog No. C118425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C118425ML 25 mL
C1184250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C118425ML Supplier TCI America Supplier No. C118425ML
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10371-45-6
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00059054
InChI Key KAKJZWSUVDRWQU-GQCTYLIASA-N
Synonym Crotonic Acid sec-Butyl Ester
PubChem CID 5366041
IUPAC Name butan-2-yl (E)-but-2-enoate
SMILES CCC(C)OC(=O)C=CC

Specifications

Color Colorless
Boiling Point 166°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Crotonate
EINECSNumber (2)-0966
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.