Learn More
Medchemexpress LLC HY-N1247 5mg Medchemexpress, Scabertopin CAS:185213-52-9 Purity:>98%
Supplier: Medchemexpress LLC HYN12475MG
Medchemexpress, HY-N1247 5mg Scabertopin CAS:185213-52-9 Scabertopin, isolated from the whole plant of Elephantopus scaber [1] , is a sesquiterpene lactone. Scabertopin has been found to be prominent anticancer constituents [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.