Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LumiraDx SARS-CoV-2 Ag Test
Shop All Roche Diagnostics Products
Click to view available options
:
SARS-CoV-2 Ag Test
Description
- Features:
- High-sensitivity, microfluidic immunofluorescence assay for direct and qualitative detection of nucleocapsid proteins in nasal swab specimens from patients suspected of COVID-19 with
- Results in 12 minutes
- Uses nasal samples to detect nucleocapsid protein antigen with 97.6 % positive agreement to an EUA approved RT-PCR method within the first 12 days of symptom onset
- For use with the LumiraDx instrument
- Includes:
- 48 LumiraDx test strips
- 48 extraction buffer vials
- 48 dropper lids
- LumiraDx SARS-CoV-2 Ag test quick reference instructions
- Notes:
- Test has not been FDA cleared or approved, but has been authorized by FDA under an EUA for use by authorized laboratories; use by laboratories certified under the CLIA, 42 U.S.C. §263a, that meet requirements to perform moderate, high or waived complexity tests
- Authorized for use at the Point of Care (POC), i.e., in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation
- Authorized only for the detection of proteins from SARS-CoV-2, not for any other viruses or pathogens
- USA only- test is authorized for duration of declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of the virus that causes COVID-19 under Section 564(b)(1) of the Federal Food, Drug, and Cosmetic Act, 21 U.S.C. § 360bbb-3(b)(1), unless the authorization is terminated or revoked sooner
Specifications
Specifications
| Control Sets | Available separately |
| Clia Complexity | Waived |
| Detectable Analytes | SARS-CoV-2 Nucleocapsid Protein Antigen |
| Type | Rapid Test |
| For Use With (Application) | Microfluidic Immunofluorescence |
| For Use With (Equipment) | LumiraDx Instrument |
| Quantity | 48 Test Strips/Pk. |
| Test Time | 12 min. |
Authorized under EUA for use by authorized labs, test has not been FDA cleared or approved
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction