Learn More
Medchemexpress LLC HY-B0343A 500mg Medchemexpress, Sarafloxacin (hydrochloride) CAS:91296-87-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0343A500MG
Encompass_Preferred
Medchemexpress, HY-B0343A 500mg Sarafloxacin (hydrochloride) CAS:91296-87-6 Sarafloxacin (hydrochloride) (A-56620 (hydrochloride)) is a quinolone antibiotic drug. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2482