Learn More
Globe Scientific Sample Cups for Abbot Architect Series Analyzers
Description
- For use with the Abbott Architect Series Analyzers
- Specifically designed for use with these products
- Polystyrene (PS) construction, natural color
Specifications
Specifications
| Color | Natural |
| Material | Polystyrene |
| Quantity | 1000 |
| Product Type | Sample Cup |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.