Learn More
Invitrogen™ UltraPure™ Salmon Sperm DNA Solution
Description
Includes
5 tubes Salmon Sperm DNA solution (1mL each, 10mg/mL)
UltraPure™ Salmon Sperm DNA Solution is a ready-to-use, sheared DNA solution that is used directly in the preparation of prehybridization and hybridization solutions.
- Prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free, RNase-free (DEPC-treated), distilled, deionized water
- DNA is sheared to average size of ≤2,000 bp
- Concentration is adjusted to 10 mg/mL
- Used to block nonspecific attachment of probe to surface of membrane
ChIP-on-Chip, Chromatin Biology, DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, RNAi, Epigenetics & Non-Coding RNA Research, Southern Blotting
Order Info
Shipping Conditions: Dry Ice
Specifications
Specifications
| Content And Storage | 5 tubes Salmon Sperm DNA solution (1 ml each, 10 mg/ml) UltraPure™ Salmon Sperm DNA is stable for 1 year at -20°C. |
| For Use With (Application) | Chromatin Biology, Nucleic Acid Gel Electrophoresis, Blotting |
| Key Functions | Block Non-Specific Hybridization |
| Product Line | UltraPure |
| Product Type | Salmon Sperm DNA Solution |
| Quantity | 5 x 1 mL |
| Reagent Type | Salmon Sperm DNA |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.