Learn More
Medicom™ SafeMask™ Premier Plus™ Earloop Mask, ASTM 2
Catalog No. 19910739 Shop All Medicom Products
Click to view available options
:
Blue
This item is not returnable. View return policy
Catalog No. 19-910-739
Supplier: Medicom™ 2040
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Moderate-barrier (ASTM Level 2) mask for examining or treating known higher-risk patients and for procedures involving moderate levels of spray and splatter.
  • Smooth, stretchy, latex-free, ultrasonically sealed earloops prevent irritation and allow easy removal with a lower risk of self-contamination.
  • Adjustable nosepiece provides a comfortable, personalized fit.
  • Shingle-pleat design prevents pooling and cross-contamination.
  • Non-woven layers will not lint, tear or shred.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 2
Color Blue
Fastening Type Earloops
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
Filter Efficiency ≥98% (PFE), ≥98% (BFE)
Latex Free Yes
Model 2040
Packaging Quantity 50 Masks/Box, 10 Boxes/Cs., 500 Masks/Cs.
Series SafeMask™ Premier Plus™
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.