Learn More
Newcomer Supply SafeCube Dispenser for Formalin 5 Gal. Cube Containment with filling tray and built in liquid detection alarm
SDP
Supplier:  Newcomer Supply ABSCD100A
Encompass
Workstation for filling containers with 10% formalin, xylene, alcohols, lab reagents, detergents, disinfectants, etc.  No more uncontrolled drips or spills. Liquid Detection Alarm in main cabinet & filling tray. Main cabinet can contain a spill up to 5 gallons. The filling tray can contain a spill up to 1/2 liter and holds up to 40 lbs. with no risk of tipping. Great for gross room, morgue, O.R., or labor and delivery. Can also be wall mounted. Dimensions: 20" (D) x 18.5" (W) x 17" (H).
Catalog No. NC9673346
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.