Learn More
Rubber Hammer
SCI_ED
Catalog No. S42065M Shop All Medivibe Technology Ltd. Products
Click to view available options
:
Large Rubber Hammer
Catalog No. S42065M
Supplier: Medivibe Technology Ltd. 700LRMLT
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Made of hard rubber and is suitable for use on tuning forks under 512Hz. Can be used safely with aluminum tuning forks.
Specifications
Product Type Rubber Hammer
Material Rubber
For Use With (Application) For exciting forks as well as resonators
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.