Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rpn2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP158214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP158214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP158214 Supplier Novus Biologicals Supplier No. NBP158214
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rpn2 Polyclonal specifically detects Rpn2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Rpn2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q99460
Gene Alias 26S proteasome non-ATPase regulatory subunit 1, 26S proteasome regulatory subunit RPN2, 26S proteasome regulatory subunit S1, 26S proteasome subunit p112, MGC133040, MGC133041, P112, proteasome (prosome, macropain) 26S subunit, non-ATPase, 1, Rpn2, S1
Gene Symbols PSMD1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PSMD1(proteasome (prosome, macropain) 26S subunit, non-ATPase, 1) The peptide sequence was selected from the N terminal of PSMD1. Peptide sequence HYTKQCVENADLPEGEKKPIDQRLEGIVNKMFQRCLDDHKYKQAIGIALE.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5707
Test Specificity This product is specific to Subunit or Isoform: 1.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.